pH electrode, low resistance, Orion™ 9162BNWP
Leverandør: Thermo Orion
9162BNWP
662-1455EA 6100 NOK
662-1455
pH electrode, low resistance, Orion™ 9162BNWP
Elektroder pH-elektroder
This low resistance electrode is ideal for use in viscous solutions.
  • Silver/silver chloride (Ag/AgCl) reference: High accuracy and broad temperature range
  • Rugged bulb, easy to clean
  • Waterproof BNC connector
  • Cap diameter: 16 mm
Warranty: 6 months from date of purchase by Buyer or 12 months from date of shipment from Seller, whichever is earlier.