Combination pH electrode, spear tip, Orion™ 8163BNWP ROSS™
Leverandør: Thermo Orion
8163BNWP
662-1502EA 8800 NOK
662-1502
Combination pH electrode, spear tip, Orion™ 8163BNWP ROSS™
Elektroder pH-elektroder
Its spear tip makes this electrode ideal for pH measurement in semi-solid food material such as cheese, meat, fruit, bread and other similar samples. Compared to conventional electrodes, the proven ROSS reference system exhibits superior stability in measurements, faster response, greater accuracy and precision when measuring samples that vary in temperature or when calibrating in temperatures that differ from the samples.
  • ROSS electrodes avoid the long term drift that other electrodes exhibit (ROSS electrodes drift by less than 0,002 pH per day), so