Combination pH electrode, Orion™ 9102BNWP
Leverandør: Thermo Orion
9102BNWP
662-1527EA 4750 NOK
662-1527
Combination pH electrode, Orion™ 9102BNWP
Elektroder pH-elektroder
This general purpose pH electrode is ideal for conducting accurate Q.C. and research work.
  • Contains a silver/silver chloride reference, widely used due to accuracy and broad temperature range
  • With waterproof connector
Leveringsinformasjon: Supplied with 60 ml bottle of Orion electrode fill solution.