Ion selective electrodes, iodide, Orion™
Leverandør: Thermo Orion
9453BN 9653BNWP 9453SC
662-0845EA 15125 NOK
662-0845 662-1574 ORIO9453SC
Ion selective electrodes, iodide, Orion™
Elektroder ISE elektroder
These electrodes are designed for measuring iodide ions in quickly, simply, accurately and economically.
  • Stable, drift-free potentials
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
  • When conventional electrode junctions clog, the Sure-Flow™ junction can be opened and flushed clean simply by pressing the cap
  • Half-cell electrode used in conjunction with the double junction reference electrode offers the benefits of the Sure-Flow™ junction design
  • Combination electrode does not require a separate reference electrode, making it