Ion selective electrodes, potassium, Orion™
Leverandør: Thermo Orion
9319BN 9719BNWP
662-0817EA 17400 NOK
662-0817 662-1578
Ion selective electrodes, potassium, Orion™
Elektroder ISE elektroder
These potassium electrodes are designed to measure potassium in aqueous solutions simply, accurately and economically.
  • Built-in Sure-Flow™ reference junction prevents electrode clogging and provides fast and stabile readings
  • Combination electrode does not require a separate reference electrode, making it convenient to use with small sample volumes
Bestillingsinformasjon: The half cell electrode requires a separate reference electrode which can be ordered separately.