Ion selective electrode, ammonia, Orion™
Leverandør: Thermo Orion
9512BNWP
662-1592EA 14550 NOK
662-1592
Ion selective electrode, ammonia, Orion™
Elektroder ISE elektroder
This standard gas-sensing ISE electrode is designed for quick, reliable and accurate measurements of dissolved ammonia in aqueous solutions.
  • Waterproof BNC connector
  • Can be used on any ISE or mV meter with a BNC connection
The ammonia combination electrode has the sensing and reference half-cells built into one electrode, which decreases the amount of required solutions and reduces waste.
Leveringsinformasjon: Electrode is supplied with 40 loose membranes, one reusable membrane cap, tweezers for handling the membranes and one bottle of electrodeMore Product Information