Ion selective electrodes, lead, Orion™
Leverandør: Thermo Orion
9482BN 9482SC 9682BNWP
662-0849EA 23625 NOK
662-0849 662-1596 662-1575
Ion selective electrodes, lead, Orion™
Elektroder ISE elektroder
These electrodes are designed for measuring lead ions in aqueous solutions quickly, simply, accurately and economically.
  • Stable, drift-free potentials
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
  • When conventional electrode junctions clog, the Sure-Flow™ junction can be opened and flushed clean simply by pressing the cap
  • Half-cell electrode used in conjunction with the double junction reference electrode offers the benefits of the Sure-Flow™ junction design
  • Combination electrode ORIO9682BNWP does not require a separate