VWR® pHenomenal® pH 1100 H pH/mV/°C Meter, Handheld
Leverandør: VWR Collection
662-1659EA 9450 NOK
662-1659
VWR® pHenomenal® pH 1100 H pH/mV/°C Meter, Handheld
Elektrokjemiske målere pH-meter
This user friendly, portable instrument with robust IP 67 rated housing provides high resolution and accuracy for precise measurements in field applications. Complete with waterproof USB output for connection to a computer. GLP compliant.
  • Large graphic LCD with continuous LED backlight shows both pH/mV and temperature value simultaneously
  • Data logging function selectable between 1 min and 1 h
  • 5000 data set memory capacity with automatic storing; additional 200-point memory for manual storage
  • Automatic power shut-off: Adjustable from 10 min to 24 h