pH/mV/°C/ion meter, handheld, pH/ION 3310
Leverandør: WTW
2GA310
662-1862EA 32425 NOK
662-1862
pH/mV/°C/ion meter, handheld, pH/ION 3310
Elektrokjemiske målere pH-meter
This professional meter with data logger is ideal for field applications. It is designed for precise pH, mV and ISE measurements.
  • Robust and waterproof
  • Backlit graphic display
  • 1- to 5-point pH calibration, 2- to 7-point ISE calibration (even non linear)
  • GLP supporting documentation
  • USB interface can be used for data transfer but also for power supply in the laboratory
Bestillingsinformasjon: Single instrument supplied with instruction manual, CD-ROM, batteries and cable. Electrodes need to be ordered separately.