ORP tester, Eutech ORPTestr 10
Leverandør: EUTECH
ORPTEST10
662-2016EA 2900 NOK
662-2016
ORP tester, Eutech ORPTestr 10
Elektrokjemiske målere pH-meter
This waterproof tester is designed with advanced microprocessor technology to give you up to ±2 mV of accuracy across a wide measuring range. User-replaceable double-junction sensor with a wide platinum band provides highly accurate results even in wet and rugged environments. Ideal for measurements in electroplating rinse tanks, swimming pools, drinking water, waste water, chemical processing, cyanide destruction and chromate reduction.
  • Large dual display for enhanced readability, provides comprehensive meter information
  • Water and dust-proof housing (IP 67