pH tester, Eutech pHTestr 30
Leverandør: EUTECH
PHTEST30
662-2017EA 2650 NOK
662-2017
pH tester, Eutech pHTestr 30
Elektrokjemiske målere pH-meter
This water- and dust-proof tester is ideal for quick and accurate pH checks in pools and spas, aquariums and hydroponics operations, cooling towers, food processing water and process/wastewater. The double junction electrode makes the tester ideal for dirty water and solutions with heavy metals or contaminants, while increased reference gel volume offers significantly longer electrode life. Also suitable for educational purposes in schools.
  • Large dual display for enhanced readability, provides comprehensive meter information
  • Water and dust-proof housing (