Multi-parameter meter, handheld, Portavo® 908 Multi
Leverandør: KNICK
908MULTI
662-2089EA 26375 NOK
662-2089
Multi-parameter meter, handheld, Portavo® 908 Multi
Elektrokjemiske målere pH-meter
This portable Memosens-based instrument for the analysis of liquids can measure pH/ORP, conductivity (conductive/inductive), oxygen (amperometric/optical) and temperature and features direct printer control. A printer for immediate printing of the calibration record (GLP compliant) can be directly connected to the micro USB port. Waterproof according to IP 66/IP 67.
  • Robust and versatile housing design with protective cover allows the meter to be used outdoors
  • Scratch-resistant, high-resolution colour graphic display with unique graphic symbols for