pH and ORP electrodes, IDS, wireless ready
Leverandør: WTW
103748 103760 103761 103762 103764 103765 103766 103767
662-1720EA 10725 NOK
662-1720 662-2049 662-2121 662-2122 662-2128 662-2129 662-2130 662-2131
pH and ORP electrodes, IDS, wireless ready
Elektroder pH-elektroder
High quality, enhanced sensor technology combined with most modern measurement electronics. The electrodes feature a plug head which can either be connected to an IDS cable or to a wireless module.
  • Intelligent: Electrodes are automatically recognized by the meter, each sensor is exactly identified
  • Digital: Measurement signals are processed digitally, providing very accurate data
  • Measurement integrity benchmarking of the electrodes by QSC (Quality Sensor Control)
  • IDS electrodes store their serial number and their calibration records
  • Wireless ready:
More Product Information