VWR® pH 110/pH 110 M, pH-/mV-/°C-måler, håndholdt

Leverandør: VWR Collection
662-1350EA 7525 NOK
662-1350 662-2268 662-2269 662-2270
VWR® pH 110/pH 110 M, pH-/mV-/°C-måler, håndholdt
Elektrokjemiske målere pH-meter
These economical, robust and waterproof pH meters are supplied in complete, ready to use kits that incorporate a robust carry case, electrode and temperature sensor with different cable lengths and buffers.

  • IP 67 waterproof housing, large display and easy to use keypad
  • Automatic/manual temperature compensation
  • 50 or 250 data sets reviewable memory
  • 1000 hour battery life; auto-shut off function (30 minutes of non use)
  • Electrode offset recognition; auto-lock and hold of readings on display

Additional functions of pH 110 M:
- Extended memory capacityMore Product Information
Bestill nå

SPESIFIKASJONER

Modell pH 110 pH 110 M pH 110 M
Environmentally Preferable
pH - område −2,00...+16,00 −2,00...+16,00
pH - oppløsning 0,01 0,01 0,01
pH - nøyaktighet ±0,01% of full scale or ±0,03 pH units ±0,01% of full scale or ±0,03 pH units
pH - kalibrering 2-point (First point must be a 6,86/7,00 buffer, the second either a 4,00/4,01 or 9,18/10,01)
Automatisk buffergjenkjenning US (4,00/7,00/10,00 @ 25 °C, 4,00/7,02/10,06 @ 20 °C)
NIST (4,01/6,86/9,18 @ 25 °C, 4,00/6,87/9,23 @ 20 °C)
US (4,00/7,00/10,00 @ 25 °C, 4,00/7,02/10,06 @ 20 °C)
NIST (4,01/6,86/9,18 @ 25 °C, 4,00/6,87/9,23 @ 20 °C)
US (4,00/7,00/10,00 @ 25 °C, 4,00/7,02/10,06 @ 20 °C)
NIST (4,01/6,86/9,18 @ 25 °C, 4,00/6,87/9,23 @ 20 °C)
mV - område ±1999 ±1999 ±1999
mV - oppløsning 1 1 1
mV - nøyaktighet ±0,1% ±1 digit ±0,1% ±1 digit
Temperaturområde −10...+120 °C −10...+120 °C −10...+120 °C
Temperaturoppløsning 0,1 °C 0,1 °C 0,1 °C
Temperaturnøyakighet ±0,3 ±1 digit ±0,3 ±1 digit
temperaturkompensering Automatic/manual (−10...+120 °C) Automatic/manual (−10...+120 °C) Automatic/manual (−10...+120 °C)
Strømforsyning One 9 Volt battery One 9 Volt battery One 9 Volt battery
Samsvar IP 67 IP 67 IP 67
Datalogg kapasitet 50 data sets 250 data sets 250 data sets
Outputs - Micro USB Micro USB
W×D×H 78×37×184 mm 78×37×184 mm 78×37×184 mm
Weight 0,272 kg 0,272 kg 0,272 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR