pH sensor with polymer body and temperature detector, SE 101 AN
Leverandør: KNICK
SE101-AN
662-2355EA 5175 NOK
662-2355
pH sensor with polymer body and temperature detector, SE 101 AN
Elektroder pH-elektroder
The SE 101-AN pH sensor features a highly robust polymer body, making it ideal for pH measurements in the fields of environmental analysis and water management.
  • Polymer electrolyte
  • Ceramic junction (2X)
  • Integrated PT 1000 temperature detector
  • Polymer body
  • pH glass with low impedance, also suitable for measurements at low temperatures