Converter Set for Lab Fermenters
Leverandør: Endress+Hauser
71537690 71537692
662-2456EA 44075 NOK
662-2456 662-2457
Converter Set for Lab Fermenters
Elektroder Elektroder med flere parametre
The CYM17 Memosens-analog converter enables easy use of digital Memosens sensors in your fermentation applications in the laboratory without changing your SOP.
  • Easy exchange of your existing analog sensors. Just install the Memosens sensors in your fermenter and connect them with the converter
  • Thanks to Memosens technology, the cable connection is insensitive to moisture, corrosion or dirt: you can count on the sensor's highest availability and your test is safe
  • Memosens sensors have an excellent performance in process applications as well as in benchtop