pH-elektroder, InLab®
Leverandør: Mettler - Toledo
51343070 51343031 51343050 51343054 51343056
662-2837EA 3980 NOK
662-2837 662-2825 662-2826 662-2830 662-2832
pH-elektroder, InLab®
Elektroder pH-elektroder
Med S7-, BNC- eller MultiPin™ (pH/temperatur)-kontakt.
  • Liquid filled electrodes for efficient pH measurements in daily laboratory use of a wide application range
  • ARGENTHAL™ reference system with silver-ion trap so that there is no risk of contamination by sulphides and proteins
  • Conform to RoHS ('Restriction of Hazardous Substances' in electrical and electronic equipment) and WEEE ('Waste Electrical and Electronic Equipment')
  • InLab® Versatile Pro features a replaceable ceramic junction which offers an economic alternative to open or movable sleeve junctions