pH electrodes, InLab® Power
Leverandør: Mettler - Toledo
51343110
662-2845EA 5980 NOK
662-2845
pH electrodes, InLab® Power
Elektroder pH-elektroder
Combination pH electrode with SteadyForce™ and S7 screw head for general laboratory applications.
  • Highly reproducible
  • Maintenance-free
  • Autoclavable
Leveringsinformasjon: Supplied with quality certificate.