VWR® pHenomenal® CO 3100 H Conductivity/TDS/°C Meter, Handheld
Leverandør: VWR Collection
663-0234EA 12900 NOK
663-0234
VWR® pHenomenal® CO 3100 H Conductivity/TDS/°C Meter, Handheld
Elektrokjemiske målere Ledningsevnemålere
This user friendly portable instrument with robust IP 67 rated housing provides high resolution and accuracy for precise measurements in field applications. Complete with waterproof USB output for connection to a computer. GLP compliant.
  • Large graphic LCD with continuous LED backlight showing conductivity, TDS and temperature values simultaneously
  • Data logging function selectable between 1 min and 1 h
  • 5000 data set memory capacity with automatic storing; additional 200 data set memory for manual storage
  • Automatic power shut-off: Adjustable from 10 min to 24