VWR® CO 310/CO 310 M, Konduktivitetsmåler, håndholdt
Leverandør: VWR Collection
663-0164EA 9225 NOK
663-0164 663-0165 663-0340 663-0341 663-0342
VWR® CO 310/CO 310 M, Konduktivitetsmåler, håndholdt
Elektrokjemiske målere Ledningsevnemålere
These portable, economical meters can simultaneously measure conductivity, specific conductance, salinity, TDS (Total Dissolved Solids) and temperature.
  • IP 67 waterproof housing, large display and easy to use keypad
  • Automatic or manual temperature compensation
  • 50 or 250 data sets reviewable memory
  • 500 (CO 310 M) or 1000 (CO 310) hour battery life; auto-shut off function (30 min of non use)
  • Electrode offset recognition; auto-lock and hold of readings on display
Additional functions of CO 310 M:
- Extended memory capacity (250 data sets)
- Waterproof USBMore Product Information