VWR® LCO 304, Conductivity Meter, Handheld
Leverandør: VWR Collection
663-0349EA 5025 NOK
663-0349
VWR® LCO 304, Conductivity Meter, Handheld
Elektrokjemiske målere Ledningsevnemålere
This high resolution device for measuring the conductivity up to 100 μS/cm is supplied with a stainless steel measuring cell for measurements in pure and ultra-pure water.
  • Measures conductivity and specific resistance
  • Permanently connected 2-pole measuring cell (stainless steel) with integrated temperature sensor
  • Dust- and waterproof (IP 65/IP 67) ABS housing
  • 3-line display with battery status indicator and backlight, protected by an unbreakable pane, overhead display at the push of a button
  • HOLD function, min./max. values, automatic measuring range