VWR® pHenomenal® IDP 761-C Conductivity Cell
Leverandør: VWR Collection
663-0356EA 7350 NOK
663-0356
VWR® pHenomenal® IDP 761-C Conductivity Cell
Elektroder Konduktivitetselektrode
This IDP (Intelligent Digital Probe) conductivity/temperature cell can be used with the pHenomenal® MD 8000 H and MD 8000 L multi-parameter meters.
  • Calibration data (serial number, calibration status) is stored in the electrode/probe, transmitted to the meter and displayed
  • Proven reliability and accuracy
  • Correct measurement results due to direct signal processing in the sensor and digital transmission of values
  • All parameters are easily measured with cable lengths of up to 100 metres
  • Same universal connector for all electrodes and probes
TemperatureMore Product Information