Dissolved oxygen probe, ProOBOD
Leverandør: YSI ENVIRONMENTAL
626401
664-0080EA 23225 NOK
664-0080
Dissolved oxygen probe, ProOBOD
Elektroder Oksygenelektrode
The ProOBOD self-stirring optical BOD probe is designed for use with the YSI ProODO and ProDSS instruments.
  • Extremely quiet operation
  • No warm up time required
  • Self-stirring to provide representative sample and increase response times
  • Smart sensor retains calibration in the probe
  • Probe body tapered to fit into a standard 300 ml BOD bottle
Bestillingsinformasjon: Supplied with optical sensor cap and power supply.