VWR® DO-220/DO-220 M, Dissolved Oxygen Meters, Handheld, with Optical sensor
Leverandør: VWR Collection
664-0168EA 6625 NOK
664-0168 664-0169 664-0170 664-0225 664-0226 664-0227
VWR® DO-220/DO-220 M, Dissolved Oxygen Meters, Handheld, with Optical sensor
Elektrokjemiske målere Målere for oppløst oksygen
These compact, portable meters measure dissolved oxygen and temperature using a smart optical sensor. Values are transmitted from and to the sensor in modified digital RS485 format. Ideal for field measurements.
  • IP 67 waterproof housing, segmented LCD showing DO and temperature values simultaneously
  • 50 or 250 data sets reviewable memory
  • Sensor cap coefficients to be manually entered into instrument by end user; sensor cap change after 12 to 16 months
  • 100 h battery life; low battery indicator; auto-shut off function (after 30 min of non use)
  • Electrode