VWR® phenomenal® OX 4100 H, Dissolved Oxygen Meter, Handheld
Leverandør: VWR Collection
664-0176EA 14500 NOK
664-0176
VWR® phenomenal® OX 4100 H, Dissolved Oxygen Meter, Handheld
Elektrokjemiske målere Målere for oppløst oksygen
This user friendly portable instrument with robust IP 67 rated housing provides high resolution and accuracy for precise measurements in field applications. Complete with waterproof USB output for connection to a computer. GLP compliant.
  • Large graphic LCD with continuous LED backlight showing both oxygen and temperature values simultaneously
  • Data logging function selectable between 1 min and 1 h
  • 5000 data set memory capacity with automatic storing; additional 200 data set memory for manual storage
  • Automatic power shut-off: Adjustable from 10 min to 24 h