Luminescent/optical dissolved oxygen (LDO) electrode for BOD measurement, IntelliCAL™ LBOD101
Leverandør: Hach
LBOD10101
664-0205EA 20150 NOK
664-0205
Luminescent/optical dissolved oxygen (LDO) electrode for BOD measurement, IntelliCAL™ LBOD101
Elektroder Oksygenelektrode
The LBOD101 is a digital, luminescent dissolved oxygen (LDO) sensor with a replaceable, integrated stirring system. The electrode measures the dissolved oxygen for BOD (Biochemical Oxygen Demand) determination in BOD bottles in BOD5 testing applications. Ideal for initial and final dissolved oxygen measurements.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • No membranes to foul or replace, no electrolyte solution to replenish, and fast, reliable 2-point calibration that stabilises quickly
  • Stirrer assembly and long-life magnetic-based motor are