VWR® DO 40, Dissolved Oxygen Meter, Handheld
Leverandør: VWR Collection
664-0234EA 4925 NOK
664-0234
VWR® DO 40, Dissolved Oxygen Meter, Handheld
Elektrokjemiske målere Målere for oppløst oksygen
This portable oxygen meter, with maintenance friendly galvanic sensor, is an entry level device suitable for everyday use. Concentrations in mg/L (ppm) and saturation in percentage can be read directly without using tables.
  • Ergonomic, dust- and waterproof ABS housing (IP 65/67)
  • 3-line display with battery status indicator and backlight, protected by an unbreakable pane, overhead display at the push of a button
  • Easy calibration at the push of a button
  • HOLD function, min./max. values, stability recognition, automatic adjustment for environmental air
  • Long