Måler for oppløst oksygen, håndholdt, Portavo 904
Leverandør: KNICK
904OXY
665-0291EA 20075 NOK
665-0291
Måler for oppløst oksygen, håndholdt, Portavo 904
Elektrokjemiske målere Målere for oppløst oksygen
This portable, intuitive meter is suitable for daily oxygen measurements. The versatile functionality and robustness makes it the perfect choice for both laboratory and process applications. Waterproof according to IP 66/IP 67.
  • Reinforced plastic enclosure for rough industrial applications
  • Integrated cover protects the display and serves as lab stand
  • Clear glass display is easy to read, even after years of use
  • Micro USB port for data connection to PC and power supply
  • With digital Memosens® input, which allows signals to be digitized and transferred