VWR® pHenomenal® MU 6100 L Multi-Parameter Meter (pH/Conductivity/DO), Bench
Leverandør: VWR Collection
665-0309EA 19300 NOK
665-0309 665-0310
VWR® pHenomenal® MU 6100 L Multi-Parameter Meter (pH/Conductivity/DO), Bench
Elektrokjemiske målere pH-meter
User friendly instrument, with articulated electrode stand and IP 43-rated housing, provides high resolution and accuracy for precise multi-parameter measurements. This meter can either measure pH/mV and conductivity or pH/mV and oxygen simultaneously. Complete with USB output for connection to a computer. GLP compliant.
  • Large LCD graphic display with continuous LED backlight shows the measured parameter and the temperature value simultaneously
  • Data logging function selectable between 1 minute and 1 hour; memory capacity of 5000 data sets