VWR® pHenomenal® MU 6100 H Multi-Parameter Meter (pH/Conductivity/DO), Handheld
Leverandør: VWR Collection
665-0311EA 17950 NOK
665-0311 665-0312
VWR® pHenomenal® MU 6100 H Multi-Parameter Meter (pH/Conductivity/DO), Handheld
Elektrokjemiske målere pH-meter
This user friendly portable instrument with robust IP 67 rated housing provides high resolution and accuracy for precise measurements of pH, mV, conductivity and oxygen in field applications (can only be used to measure any two of these at the same time). Complete with waterproof USB output for connection to a computer. GLP compliant.
  • Large LCD graphic display with continuous LED backlight showing the measured parameter and the temperature value simultaneously
  • Data logging function selectable between 1 min and 1 H
  • 5000 data set memory capacity with