Puncture pH combination electrode, IntelliCAL™ PHC108
Leverandør: Hach
PHC10801
665-0383EA 6100 NOK
665-0383
Puncture pH combination electrode, IntelliCAL™ PHC108
Elektroder pH-elektroder
Robust digital puncture pH electrode with stainless steel body, Ag/AgCl reference system and built-in temperature sensor.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Digital electrode stores time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number
  • Can be moved between meters without the need to re-calibrate or re-enter measurement settings
  • Ideal for a multi-user environment with multiple HQD series laboratory and portable meters
  • Ultimate traceability in measurement history
  • Non