VWR® pH CO 1030, pH/Conductivity Tester
Leverandør: VWR Collection
665-0480EA 3100 NOK
665-0480
VWR® pH CO 1030, pH/Conductivity Tester
Elektrokjemiske målere pH-meter
This tester is a convenient pen instrument for measuring the following water quality parameters: pH, conductivity, TDS, salinity, and temperature.
  • Graphic LCD, IP 67 waterproof housing
  • Replaceable electrode
  • Automatic temperature compensation; automatic calibration and buffer recognition
  • HOLD function, battery indicator, automatic power off function after 10 minutes of non use
  • Non volatile memory stores up to 50 sets of data (pH/conductivity/TDS/salinity and temperature) even if power is lost
  • Belt clip for easy storage when not in use
Warranty: 1More Product Information