VWR® pHenomenal® MD 8000 L Bench Top Digital Multi-Parameter Instruments
Leverandør: VWR Collection
665-0499EA 16600 NOK
665-0499 665-0498
VWR® pHenomenal® MD 8000 L Bench Top Digital Multi-Parameter Instruments
Elektrokjemiske målere pH-meter
These digital meters combine six parameters in one instrument: pH, mV, temperature, conductivity, dissolved oxygen and turbidity. IDP technology optimises use and makes measurement easier (no need to recalibrate when changing the meter, and no need to adjust the meter's settings). The measured data is complete with serial number, settings and calibration data.
  • One universal measuring channel
  • Digital sensor recognition: Measured parameter and settings are recognised automatically
  • Suitable for Intelligent Digital Probes (IDP)
  • Suitable for measurements with