VWR® pH CO 2500L, pH/Conductivity/°C Bench Meter with Colour Graphic Touch Screen
Leverandør: VWR Collection
665-0561EA 13650 NOK
665-0561
VWR® pH CO 2500L, pH/Conductivity/°C Bench Meter with Colour Graphic Touch Screen
Elektrokjemiske målere pH-meter
User friendly instrument distinguished by a large a 12,7 cm colour graphic touch screen. Can be used for accurate measurements of pH, mV, Redox potential (ORP), conductivity, salinity, TDS, resistivity and temperature. This pH meter enables simultaneous measurements displaying and storing of three functions (pH, conductivity and temperature).
  • Large five colour graphic touch screen showing pH/mV, conductivity and temperature value simultaneously
  • 1- to 3-point calibration with automatic detection of buffer solutions and NIST standard solutions
  • Storing