Supelco® BIOshell™ Guard Cartridge Holder
Leverandør: Merck
66841-U
66841-UEA 4025 NOK
66841-U
Supelco® BIOshell™ Guard Cartridge Holder
Kolonneholdere
This guard cartridge holder can be used with 4,6/3,0 and 2,1 mm Int.Ø BIOShell™ columns.