Petroleumbensin, 40…60 °C, AR®, Laboratory Reagent, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger

6722-45EA 6400 NOK
6722-45 6722-25
Petroleumbensin, 40…60 °C, AR®, Laboratory Reagent, Macron Fine Chemicals™
Petroleumbensin
MDL nummer: MFCD00081849
CAS-nummer: 64742-49-0
FN: 1268
ADR: 3,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR