Pyridin ≥99.5%, ChromAR® for HPLC, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger

Synonymer: Azabenzene, Azine

6730-25EA 4350 NOK
6730-25
Pyridin ≥99.5%, ChromAR® for HPLC, Macron Fine Chemicals™
Pyridine

Suitable for Liquid Chromatography and UV-Spectrophotometry

Formel: C₅H₅N
MW: 79.1 g/mol
Kokepunkt: 115.5 °C
Smeltepunkt: -44
Tetthet: 0.98
Flash punkt: 17 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011732
CAS-nummer: 110-86-1
FN: 1486
ADR: 3
Merck Indeks: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (GC) min. 99.5 %
Residue after Evaporation max. 0.005 %
Water (KF) max. 0.05 %
Ultraviolet Absorbance (1.00-cm cell vs water):
at 340 nm max. 0.07
at 350 nm max. 0.02
at 400 nm max. 0.01

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR