Kaliumhydrogenkarbonat 99.7-100.5%, granular, AR® ACS, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials


Management of Change (MOC) category = N

6748-20EA 0 NOK
6748-20
Kaliumhydrogenkarbonat 99.7-100.5%, granular, AR® ACS, Macron Fine Chemicals™
Kaliumhydrogenkarbonat
Formel: KHCO₃
Smeltepunkt: 292 °C
Tetthet: 2,17 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011402
CAS-nummer: 298-14-6
EINECS: 206-059-0
Merck Indeks: 13,07691

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use 99.7 - 100.5 %
ACS - Assay (KHCO₃) 99.7 - 100.5 %
ACS - Ammonium (NH₄) ≤0.0005 %
ACS - Calcium (Ca) ≤0.002 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤0.0005 %
ACS - Insoluble Matter ≤0.01 %
ACS - Iron (Fe) ≤5 ppm
ACS - Magnesium (Mg) ≤0.001 %
ACS - Phosphate (PO₄) ≤0.0005 %
ACS - Sodium (Na) ≤0.03 %
ACS - Sulfur Compounds (as SO₄) ≤0.003 %
Appearance (colorless, transparent granules)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR