Hexan fraksjon, ChromAR® for HPLC, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger

6751-25EA 3150 NOK
6751-25
Hexan fraksjon, ChromAR® for HPLC, Macron Fine Chemicals™
Hexan fraksjon

Suitable for Liquid Chromatography and UV-Spectrophotometry

Kokepunkt: 69 °C (1013 hPa)
Tetthet: 0,659 g/cm³ (20 °C)
Flash punkt: –22 °C
Lagringstemperatur: Romtemperatur
CAS-nummer: 110-54-3
FN: 1208
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (GC) min. 45 %
Residue after Evaporation max. 0.0005 %
Titrable Acid (meq/g) max. 0.0005
Water (KF) max. 0.01 %
Ultraviolet Absorbance (1.00-cm cell vs water):
at 210 nm max. 0.2
at 220 nm max. 0.1
at 230 nm max. 0.02
at 240 nm max. 0.01
at 250 nm max. 0.004


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR