n-Pentan ≥95%, ChromAR® for HPLC, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger

6755-25EA 2775 NOK
6755-25
n-Pentan ≥95%, ChromAR® for HPLC, Macron Fine Chemicals™
n-Pentane

Suitable for Liquid Chromatography and UV-Spectrophotometry

Formel: H₃C(CH₂)₃CH₃
Kokepunkt: 36,1 °C
Tetthet: 0,63
Flash punkt: –48 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009498
CAS-nummer: 109-66-0
FN: 1265
ADR: 3,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (GC) min. 95 %
Residue after Evaporation max. 0.0005 %
Titrable Acid (meq/g) max. 0.0005
Water (KF) max. 0.01 %
Ultraviolet Absorbance (1.00-cm cell vs water):
at 210 nm max. 0.4
at 220 nm max. 0.15
at 230 nm max. 0.05
at 240 nm max. 0.01
at 250 nm max. 0.004


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR