2-Decanon, Supelco®

Leverandør: Merck

Synonymer: Decan-2-one

M-Clarity™ quality level = MQ100

68228-5ML
68228-5MLEA 883 NOK
68228-5ML
2-Decanon, Supelco®
2-Decanone
Formel: C₁₀H₂₀O
MW: 156,27 g/mol
Kokepunkt: 211 °C (1013 hPa)
Smeltepunkt: 3 °C
Tetthet: 0,82 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009571
CAS-nummer: 693-54-9
EINECS: 211-752-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2541741e82c2ff2a0284c203d448f9155c1a1d4&vid=c3dea62c240de13e76072926a1b0085eb1856fe3&prid=web&clid=SAPDAM

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR