2-Metylpentan, Supelco®

Leverandør: Merck
Danger

Synonymer: 1,1-Dimethylbutane, Isohexane

M-Clarity™ quality level = MQ100

68310-50ML
68310-50MLEA 4875 NOK
68310-50ML
2-Metylpentan, Supelco®
2-Metylpentan
Formel: C₆H₁₄
MW: 86.18 g/mol
Kokepunkt: 62
Smeltepunkt: –154 °C
Tetthet: 0.653 g/cm³ (20 °C)
Flash punkt: –33 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009406
CAS-nummer: 107-83-5
EINECS: 203-523-4
FN: 1208
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f6345e7519094f39e2f0ebebd0ff76e9c4bcb74&vid=0bcf9819f50ad290ac95864472c28e199424a43c&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR