Supelco® Organic Reference Standard, cis-8,11,14-Eicosatrienoic acid

Leverandør: Merck

69154-50MG
69154-50MGEA 1650 NOK
69154-50MG
Supelco® Organic Reference Standard, cis-8,11,14-Eicosatrienoic acid
Standarder Organiske standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=130e6899f61810ec3f1df88e3c7fbf15071b4a6c&vid=130e6899f61810ec3f1df88e3c7fbf15071b4a6c&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Synonym(s) Homo-γ-linolenic acid
M-Clarity quality level MQ100
CAS-nr 1783-84-2
MDL No. MFCD00065721
Formel CH₃(CH₂)₃(CH₂CH=CH)₃(CH₂)₆CO₂H
Molekylvekt 306,48
Flammepunkt 62 °C - Closed cup
Fraktforhold WICE
Lagringsforhold –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR