Zink 1000 µg/ml, BAKER INSTRA-ANALYZED® for spormetallanalyse, J.T.Baker®

Leverandør: Avantor
Warning

6946.0100EA 0 NOK
6946.0100
Zink 1000 µg/ml, BAKER INSTRA-ANALYZED® for spormetallanalyse, J.T.Baker®
Zinc
Formel: Zn
MDL nummer: MFCD00011291
CAS-nummer: 7440-66-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR