Organic reference standard, 3-Ethylmalic acid, ≥95,0% (HPLC), Supelco®

Leverandør: Merck

69474-10MG
69474-10MGEA 1500 NOK
69474-10MG
Organic reference standard, 3-Ethylmalic acid, ≥95,0% (HPLC), Supelco®
Standarder Organiske standarder

Metabolite glyoxalate-condensing enzyme of human pathogens.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8934517b118f7d16a4e47bf4c94cd8d3ab117d7d&vid=8934517b118f7d16a4e47bf4c94cd8d3ab117d7d&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Beskrivelse 3-Ethylmalic acid
Environmentally Preferable
CAS-nr 16048-76-3
Formel C₆H₁₀O₅
Lagringstemp 2...8°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR