VWR®, Gel Cutters and Safety Razors
Leverandør: VWR Collection
700-0417EA 395 NOK
700-0417 700-0418
VWR®, Gel Cutters and Safety Razors
Barberkniver
Made from food grade PET. At just 0,27 mm, the VWR® gel cutter is as thin as a single edge industrial razor blade, but much safer to use.
  • Able to slice gels directly on the light box, without scratching the surface of the box*
  • Can be used to chop, mince or slice soft tissue samples
  • Adjustable - Simply cut it with standard scissors to the required width
  • Available in two sizes: Regular, sized the same as a single edge industrial razor blade, and large (30% larger)
* Best for 1 to 2% gels.