VWR® Midi S PerfectBlue, Gel system

Leverandør: VWR Collection
700-0814EA 15250 NOK
700-0814
VWR® Midi S PerfectBlue, Gel system
Elektroforesesystemer Horisontal gelelektroforesesystemer
Capable of rapidly running up to 160 samples, the Midi S is a compact, but highly efficient, electrophoresis system. Versatility is built-in, with six different comb positions and a choice of 10 different comb types and thicknesses available. For rapid handling, special microtitre combs enable rapid loading with a multichannel pipette, while the effective rubber sealed 'end gates' allow gel pouring right in the tray. With its sturdy design and straight electrodes, the results are clear, crisp and quick, free from 'smiling' bands.

*Based on an agarose gel of approximately 5 mm thickness

Bestill nå

SPESIFIKASJONER

Modell Midi S
Environmentally Preferable
Gelstørrelse B × L 150×150 mm
Prøvekapasitet 10 – 160
Gelvolum ~120 ml
Buffervolum ~1000 ml
Kjøretid ≥45 h
Enhet, B × L × H 215×275×100 mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR