VWR® Mini, Power Supply
Leverandør: VWR Collection
700-0856EA 3600 NOK
700-0856
VWR® Mini, Power Supply
Strømkilder
VWR® mini power supply is a basic, economical unit perfect for small nucleic acid gel tanks.
  • Compact and simple to operate
  • Ideal for basic agarose nucleic acid applications
  • Two pairs of output jacks
The user can select either 100 or 200 V output. All common horizontal gel tanks are compatible with the standard output jacks.