VWR® PerfectBlue Maxi S Plus, Gel system
Leverandør: VWR Collection
700-0876EA 14275 NOK
700-0876
VWR® PerfectBlue Maxi S Plus, Gel system
Elektroforesesystemer Horisontal gelelektroforesesystemer
The smallest of the Maxi series, the Maxi S Plus is suitable for high quality sample separation of up to 160 samples per run. With this flexible system, the gel size can be split into two, three or four sections by using different comb positions, plus the gel size can be reduced using the wall comb or the casting dam system.
The system is supplied with three combs but others are available – either standard combs, or microtitre combs which are ideal for use with multichannel pipettes. Use of the system end gates ensure that gel pouring is leak-free and easy. The levelling table keeps theMore Product Information