VWR® PerfectBlue Maxi M 'Revolution', Gel system
Leverandør: VWR Collection
700-0909EA 22250 NOK
700-0909
VWR® PerfectBlue Maxi M 'Revolution', Gel system
Elektroforesesystemer Horisontal gelelektroforesesystemer
Based on the Maxi M model, the Maxi M 'Revolution' offers a built-in buffer circulation system, powered by the system’s electrodes. At the cathode, hydrogen bubbles are produced and pass up a diagonally positioned pipe connected to the two sides of the tank to produce a gentle and continual circulation of the buffer. The system nulls any pH and ionic gradient – without the fuss of mechanical pumps. The system offers 6 comb positions enabling the gel to be partitioned into 2, 3 or 4 areas. In addition, the system can accommodate any of the 23 standard and microtitre comb designs described forMore Product Information
Bestill nå
SPESIFIKASJONER
Modell Maxi M 'Revolution'
Gelstørrelse B × L 200×250 mm
Environmentally Preferable
Prøvekapasitet 8 – 168
Gelvolum 250 ml
Buffervolum ~2000 ml
Kjøretid ≥60 h
Enhet, B × L × H 270×380×155 mm